Order Entry
Czech Republic
ContactUsLinkComponent
 
Organic reference standard, Diethyl phthalate 100 µg/ml in n-nonane, CIL
  PROOCILULM61741.2
 :  
Organic reference standard, Diethyl phthalate 100 µg/ml in n-nonane, CIL
  PROOCILULM61741.2
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    84-66-2
  • Description:
    Diethyl phthalate 100 µg/ml in n-nonane
  • Formula:
    C₁₂H₁₄O₄
  • Description:
    Diethyl phthalate