Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1.013 hPa)
Melting Pt:6.4 °C
Density:0.78 g/cm³ (20 °C)
Flash Pt:8.3 %(V)
EINECS:203-806-2
UN:1145
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:complying
- Assay (GC):Min. 99.5 %
- Solidification range:5.5 - 6.5 °C
- Boiling range:79 - 81 °C
- Vapor Density:2.9 (vs air)
- Transition Temp:solidification point 5.5-6.5 °C
- Density (D 20/4):0.778 - 0.779
- Refractive index (n 20/D):1.4250 - 1.4280
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as HCl):Max. 0.001 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- Aromatic substances (as C₆H₆):Max. 0.05 %
- cyclohexene (C₆H₁₀):Max. 0.05 %
- Reaction against H₂SO₄:complying
- APHA:Max. 10
Specification Test Results
Frequently Bought Together