Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-3-bromo-5-hydroxybenzoate ≥97%
Irritant
Methyl-3-bromo-5-hydroxybenzoate ≥97%
Catalog # H51724.03
CAS Number:  
Irritant
Methyl-3-bromo-5-hydroxybenzoate ≥97%
Catalog # H51724.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :231.05 g/mol
Melting Pt:130…135 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:192810-12-1
Formula:C₈H₇BrO₃
MDL Number:MFCD10566800