


2-Chloroethyldiisopropylammonium chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4261-68-1
:
Formula:C₈H₁₉Cl₂N
MDL Number:MFCD00012496
:200.15 g/mol
Melting Pt:131…134 °C
Storage Temperature:Ambient
EINECS:224-238-1
UN:2811
ADR:6.1,I
:GHS06
:GHS05
:GHS07
Related Information
Frequently Bought Together