
Pyridinium-p-toluenesulphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24057-28-1
:
Formula:C₁₂H₁₃NO₃S
MDL Number:MFCD00013108
:251.31 g/mol
Melting Pt:116…120 °C
EINECS:246-002-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together