2-Chloroethyldiethylammonium chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Diethylaminoethylchloride hydrochloride, N-(2-Chloroethyl)-N,N-diethylammonium chloride
CAS Number:869-24-9
:
Formula:C₆H₁₅Cl₂N
MDL Number:MFCD00012519
:172.1 g/mol
Melting Pt:206…210 °C
Storage Temperature:Ambient
EINECS:212-786-4
UN:2928
ADR:6.1,II
:GHS06
:GHS05
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together