Order Entry
Czech Republic
ContactUsLinkComponent
Menthone (mixture of isomers) 98%
Menthone (mixture of isomers) 98%
Catalog # ACRO125411000
CAS Number:  
undefined
Menthone (mixture of isomers) 98%
Catalog # ACRO125411000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4460 to 1.4550 (20°C, 589 nm)
  • Density:
    (20°C) 0.889 to 0.898 g/ml
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :154.25 g/mol
Melting Pt:–6 °C
Density:0.89 g/cm³
Flash Pt:69 °C
Storage Temperature:Ambient
EINECS:233-944-9
CAS Number:10458-14-7
Formula:C₁₀H₁₈O
MDL Number:MFCD00062998