Order Entry
Czech Republic
ContactUsLinkComponent
Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®
Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Cyclohexane, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay (by GC):
    min. 99.0 %
  • Color (APHA):
    max. 10
  • Residue after Evaporation:
    max. 0.002 %
  • Substances Darkened by H2SO4:
    passes test
  • Water (H2O):
    max. 0.02 %
  • at 210 nm:
    max. 1.00
  • at 220 nm:
    max. 0.50
  • at 230 nm:
    max. 0.20
  • at 240 nm:
    max. 0.08
  • at 250 nm:
    max. 0.03
  • at 260 nm:
    max. 0.02
  • at 300-400 nm:
    max. 0.01

Specification Test Results

Related Information

Frequently Bought Together