Order Entry
Czech Republic
ContactUsLinkComponent
 
1,4-Cyclohexanedione 98%
 
undefined
1,4-Cyclohexanedione 98%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Yellow to beige Powder or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    76 to 80 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together