Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Cyclohexanone, Supelco®

Glass bottle, 100 mL
Catalog # 29135-100ML
Flammable
Irritant
Supplier:   Merck
Cyclohexanone, Supelco®
Catalog # 29135-100ML
Supplier:  Merck
CAS Number:  108-94-1
Cyclohexanone, Supelco®
Cyclohexanone, Supelco®

Glass bottle, 100 mL

CAS Number:  108-94-1
Price
2 380,00 Kč
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications