Order Entry
Czech Republic
ContactUsLinkComponent
p-Xylene ≥99%, GPR RECTAPUR®
Flammable
Irritant
p-Xylene ≥99%, GPR RECTAPUR®
Catalog # 28984.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
p-Xylene ≥99%, GPR RECTAPUR®
Catalog # 28984.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Boiling point:
    137 - 139 °C
  • Solidification point:
    12 - 14 °C
  • Refractive index (20°C):
    1.495 - 1.499
  • Evaporation residue:
    Max. 50 ppm

Specification Test Results

About this item

 :
 :106.17 g/mol
Merck Index:12,10214
Boiling Pt:138.3 °C (1013 hPa)
Melting Pt:14 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:203-396-5
UN:1307
ADR:3,III
 :GHS02
 :GHS07
CAS Number:106-42-3
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00008556