Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
2,6-Diisopropylaniline for synthesis, Sigma-Aldrich®
Catalog # 8.41095.9999
Supplier:  Merck
CAS Number:  
Irritant
2,6-Diisopropylaniline for synthesis, Sigma-Aldrich®
Catalog # 8.41095.9999
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Solubility:
    < - 0.2 g/l (20 °C)
  • Melting Point:
    -45 °C
  • Molar Mass:
    177.29 g/mol
  • Boiling Point:
    257 °C (1013 hPa)
  • Flash Point:
    125 °C
  • Density:
    0.94 g/cm3 (20 °C)
  • Ignition Point:
    400 °C DIN 51794

Specification Test Results

About this item

 :
 :177.29 g/mol
Boiling Pt:257 °C (1013 hPa)
Melting Pt:–45 °C
Density:0.94 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:246-305-4
 :GHS07
CAS Number:24544-04-5
Formula:C₁₂H₁₉N
MDL Number:MFCD00008887