Order Entry
Czech Republic
ContactUsLinkComponent
 
Urea 8 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALU4883-6X25ML
Supplier:  Merck
CAS Number:  
undefined
Urea 8 mol/l in aqueous solution, Sigma-Aldrich®
Catalog # SIALU4883-6X25ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    150 mL
  • Seq:
    2500

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
 :60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
CAS Number:57-13-6
Formula:NH₂CONH₂
MDL Number:MFCD00008022