
Hydrochloric acid 1 mol/l (1 N), Sigma-Aldrich®
Catalog # SIALH5409-3.8L
Supplier: Merck
CAS Number:
Specifications
- Pk:3,8 L
- Seq:5138
Specifications
About this item
:
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324