N-(2-Hydroxyethyl)morpholine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:622-40-2
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00006180
:131.17 g/mol
:M-Clarity™ quality level = MQ200
EINECS:210-734-5
Frequently Bought Together