3-Chlorophenethyl bromide, Sigma-Aldrich®
Catalog # SIAL655058-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:219.51 g/mol
CAS Number:16799-05-6
Formula:C₈H₈BrCl
MDL Number:MFCD04117465