2-Bromophenethylamine, Sigma-Aldrich®
Catalog # SIAL534099-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
:200.08 g/mol
CAS Number:65185-58-2
Formula:C₈H₁₀BrN
MDL Number:MFCD01529874