Tetrabutylammonium sulphate 50% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2472-88-0
:
Formula:C₃₂H₇₂N₂O₄S
MDL Number:MFCD00216632
:581 g/mol
:M-Clarity™ quality level = MQ200
EINECS:219-593-4
Frequently Bought Together