Trimethylaluminium 2 mol/l in heptane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-24-1
:
Formula:C₃H₉Al
MDL Number:MFCD00008252
:72.09 g/mol
ADR:4.2,I
:M-Clarity™ quality level = MQ100
EINECS:200-853-0
UN:3394
Frequently Bought Together