
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane, Sigma-Aldrich®
Catalog # SIAL282316-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:400.06 g/mol
:M-Clarity™ quality level = MQ100
EINECS:206-386-9
:GHS07
CAS Number:335-27-3
Formula:C₈F₁₆
MDL Number:MFCD00001469