Benzyltrimethylammonium hydroxide 40% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-85-6
:
Formula:C₁₀H₁₇NO
MDL Number:MFCD00008281
:167.25
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:202-895-5
UN:3267
Frequently Bought Together