2-Chlorophenethylamine, Sigma-Aldrich®
Catalog # SIAL243736-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:155.63 g/mol
:M-Clarity™ quality level = MQ100
EINECS:235-982-1
CAS Number:13078-80-3
Formula:C₈H₁₀ClN
MDL Number:MFCD00008185