Tetrabutylammonium fluoride 1 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:429-41-4
:
Formula:C₁₆H₃₆FN
MDL Number:MFCD00011747
ADR:3,II
:M-Clarity™ quality level = MQ200
:261.47 g/mol
EINECS:207-057-2
UN:2924
Frequently Bought Together