Order Entry
Czech Republic
ContactUsLinkComponent
Lidocaine hydrochloride
Lidocaine hydrochloride
Catalog # PROOICRS0338
CAS Number:  
Lidocaine hydrochloride
Catalog # PROOICRS0338
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
EINECS:200-803-8
UN:2811
ADR:6.1,III
CAS Number:73-78-9
Formula:C₁₄H₂₃ClN₂O