Organic reference standard, Sorbic acid 10 µg/ml in methanol
Catalog # PROONE9322
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:110-44-1
- Description:Sorbic acid 10 µg/ml in methanol
- Formula:C₆H₈O₂
- Description:Sorbic acid