Lidocaine hydrochloride
Catalog # PROOBP214
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
EINECS:200-803-8
UN:2811
ADR:6.1,III
CAS Number:73-78-9
Formula:C₁₄H₂₃ClN₂O