

Farnesol (mixture of isomers), clear, colourless liquid
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:4602-84-0
:
Formula:C₁₅H₂₆O
MDL Number:MFCD00002918
:222.37 g/mol
Storage Temperature:Freezer
EINECS:225-004-1
UN:3082
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together