Order Entry
Czech Republic
ContactUsLinkComponent
 
Pesticide reference standard, 2-Chloronaphthalene 100 µg/ml in n-nonane, CIL
  PROOCIL-ECN-2611
 :  
Pesticide reference standard, 2-Chloronaphthalene 100 µg/ml in n-nonane, CIL
  PROOCIL-ECN-2611
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    91-58-7
  • Description:
    2-Chloronaphthalene 100 µg/ml in n-nonane
  • Formula:
    C₁₀H₇Cl
  • Description:
    2-Chloronaphthalene