Pesticide reference standard, 1-Chloronaphthalene 100 µg/ml in n-nonane, CIL
Catalog # PROOCIL-ECN-2610
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:90-13-1
- Description:1-Chloronaphthalene 100 µg/ml in n-nonane
- Formula:C₁₀H₇Cl
- Description:1-Chloronaphthalene