
(S)-(-)-4-Fluoro-α-methylbenzylamine 99%, ee 99%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:66399-30-2
:
Formula:C₈H₁₀FN
MDL Number:MFCD03092999
:139.17 g/mol
Boiling Pt:76 °C (22 mmHg)
Melting Pt:–30 °C
Density:1.030
Flash Pt:68 °C (154 °F)
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
Related Information
Frequently Bought Together