
trans,trans-Farnesol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-28-5
:
Formula:C₁₅H₂₆O
MDL Number:MFCD00002918
:222.37 g/mol
Merck Index:14,3937
Boiling Pt:137 °C (3 mmHg)
Density:0.886
Flash Pt:96 °C (204 °F)
EINECS:225-004-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together