

4-(Dimethylamino)benzonitrile 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1197-19-9
:
Formula:C₉H₁₀N₂
MDL Number:MFCD00001815
Boiling Pt:317…319 °C
Melting Pt:72…76 °C
Storage Temperature:Ambient
EINECS:214-819-8
UN:3439
ADR:6.1,III
:146.19 g/mol
:GHS06
:GHS07
Related Information
Frequently Bought Together