
Acetophenone oxime 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:613-91-2
:
Formula:C₈H₉NO
MDL Number:MFCD00013931
:135.17 g/mol
Boiling Pt:118…120 °C (20 mmHg)
Melting Pt:58…60 °C
EINECS:210-360-2
:GHS07
Related Information
Frequently Bought Together