Boron nitride, rods, Length: 300 mm, Ø 6.4 mm
Katalogové číslo 45850.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:24.82 g/mol
Merck Index:13,01336
Melting Pt:2700 °C
Density:2.25
Storage Temperature:Ambient
EINECS:233-136-6
CAS Number:10043-11-5
Formula:BN
MDL Number:MFCD00011317