Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
 
2-Butanone oxime 99%
 
undefined
 
 
 
 
2-Butanone oxime 99%

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4410 to 1.4440 (20°C, 589 nm)
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together