Triethylaluminium 1.3 mol/l in heptane, AcroSeal®
:97-93-8
:
:C₆H₁₅Al
:MFCD00009015
:114.17 g/mol
:–93 °C
:202-619-3
:3394
:GHS07
:GHS08
:GHS05
:GHS09
:GHS02
- Appearance:Clear colorless Solution
- Strength:19.5 to 20.5 wt % (on Al basis)
Frequently Bought Together