ContactUsLinkComponent
Zinc chloride 1.0 M in diethyl ether, AcroSeal®
Irritant
Corrosive
Environment
Flammable
Zinc chloride 1.0 M in diethyl ether, AcroSeal®
Certificates
undefined
Irritant
Corrosive
Environment
Flammable
Zinc chloride 1.0 M in diethyl ether, AcroSeal®
Specification Test Results
  • Appearance:
    Colorless to light yellow to light blue-grey Solution (May form two phases which can recombine on cooling. May develop some turbidity or precipitate)
  • Titration Argentometric:
    0.9 to 1.1 M
  • Water:
    ≤0.2 % (Coulometric)