Order Entry
Czech Republic
ContactUsLinkComponent
2-Chlorophenethylamine 97%
Corrosive
2-Chlorophenethylamine 97%
Catalog # ACRO222840100
CAS Number:  
undefined
Corrosive
2-Chlorophenethylamine 97%
Catalog # ACRO222840100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5500 to 1.5530 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :155.63 g/mol
Boiling Pt:120 °C (15 mmHg)
Flash Pt:108 °C
Storage Temperature:Ambient
EINECS:235-982-1
UN:2735
ADR:8,III
 :GHS05
CAS Number:13078-80-3
Formula:C₈H₁₀ClN
MDL Number:MFCD00008185