Order Entry
Czech Republic
ContactUsLinkComponent
N-(2-Hydroxyethyl)morpholine 99%
Irritant
N-(2-Hydroxyethyl)morpholine 99%
Certificates
undefined
Irritant
N-(2-Hydroxyethyl)morpholine 99%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4760 to 1.4780 (20°C, 589 nm)
  • Water:
    ≤0.5 %
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together