Order Entry
Czech Republic
ContactUsLinkComponent
Chromium ≥99.8%, chips
Chromium ≥99.8%, chips
Catalog # ABCRAB119337.0050
Supplier:  abcr
CAS Number:  
Chromium ≥99.8%, chips
Catalog # ABCRAB119337.0050
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

About this item

 :
Merck Index:12,02288
Boiling Pt:2200 °C
Melting Pt:1890 °C
Density:7.20
 :52 g/mol
Storage Temperature:Ambient
EINECS:231-157-5
CAS Number:7440-47-3
Formula:Cr
MDL Number:MFCD00010944