

Trityl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-83-5
:
Formula:C₁₉H₁₅Cl
MDL Number:MFCD00000813
:278.78 g/mol
Boiling Pt:230…235 °C (20 mmHg)
Melting Pt:110…113 °C
Density:1,25 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-986-4
UN:3261
ADR:8,II
:GHS05
:GHS09
Specification Test Results
- Appearance:White to off-white to yellow to light beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:109 to 113 °C
- Titration Argentometric:≥97.5 % (ex Cl)
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together