Order Entry
Czech Republic
ContactUsLinkComponent
α-Phenylbenzyl bromide 90%
Corrosive
Irritant
α-Phenylbenzyl bromide 90%
Certificates
undefined
Corrosive
Irritant
α-Phenylbenzyl bromide 90%

Specification Test Results

  • Appearance:
    Orange to brown Low melting crystalline mass or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    33 to 42 °C
  • GC:
    ≥88 %

Specification Test Results

Related Information

Frequently Bought Together