
meso-1,2-Dibromo-1,2-diphenylethane ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13440-24-9
:
Formula:C₁₄H₁₂Br₂
MDL Number:MFCD00000137
Melting Pt:∼ 241 °C (dec.)
EINECS:227-324-7
ADR:8,III
:340.07 g/mol
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together