ContactUsLinkComponent
Grade 8 ruled filter papers, Whatman™
Grade 8 ruled filter papers, Whatman™
Certificates
undefined
Grade 8 ruled filter papers, Whatman™
Specifications
  • Catalog No:
  • WHAT10347008
  • WHAT10347004
  • WHAT10347033
  • WHAT10347009
  • Weight:
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • 65 g/m²
  • Particle retention:
  • 20 - 30 µm
  • 20 - 30 µm
  • 20 - 30 µm
  • 20 - 30 µm