Order Entry
Czech Republic
ContactUsLinkComponent
Toluene, Uvasol® for spectroscopy, Supelco®
Toluene, Uvasol® for spectroscopy, Supelco®
Catalog # 1.08331.1000
Supplier:  Merck
CAS Number:  
undefined
Toluene, Uvasol® for spectroscopy, Supelco®
Catalog # 1.08331.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    0.52 g/L (20 °C)
  • Melting Point:
    -95 °C
  • Molar Mass:
    92.14 g/mol
  • Boiling Point:
    110.6 °C (1013 hPa)
  • Vapor Pressure:
    29 hPa (20 °C)
  • Flash Point:
    4 °C
  • Refractive Index:
    1.4968 (20 °C)
  • Explosion Limit:
    1.2 - 8%(V)
  • Density:
    0.87 g/cm³ (20 °C)
  • pH:
    (H₂O) not applicable
  • Ignition Point:
    535 °C
  • Viscosity Kinematic:
    0.7 mm²/s (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

About this item

CAS Number:108-88-3
 :
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
 :92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
 :M-Clarity™ quality level = MQ100
Flash Pt:4 °C
REACH:05-2114615130-69
 :GHS02
 :GHS08
 :GHS07