ContactUsLinkComponent
Tubing cutter
Tubing cutter
  BURK0006-9506
 :  BURKLE
undefined
Tubing cutter
  BURK0006-9506
 :  BURKLE
  • Description:
    Plastic tubing cutter for flexible tubing from 1,6 to 19 mm diameter
  • Pk:
    1
Edges cut with this tubing cutter are always clean and flat, even in the case of thick tubing.