


Toluene
Supplier: EHRENSTORFER
Certificates
About this item
CAS Number:108-88-3
:
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS02
:GHS08
:GHS07
Frequently Bought Together