Toluene ≥99%
: Methylbenzene, Phenylmethane, FM approval
:108-88-3
:
:C₆H₅CH₃
:MFCD00008512
:111 °C
:∼–93 °C
:0.867
:92.14 g/mol
:4 °C (40 °F)
:1294
:3 II
:13,09607
:Ambient
:203-625-9
:05-2114615130-69
:GHS02
:GHS08
:GHS07
- Environmentally Preferable:
Frequently Bought Together