Ethylbenzene reference substance for gas chromatography, Supelco®
1.09635.0005
: Merck
:
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
- Solubility:0.2 g/l (20 °C)
- Melting Point:-95 °C
- Molar Mass:106.16 g/mol
- Boiling Point:136 °C (1013 hPa)
- Vapor Pressure:9.5 hPa (20 °C)
- Flash Point:15 °C
- Explosion Limit:1.0 - 7.8 %(V)
- Density:0.87 g/cm3 (20 °C)
- Ignition Point:430 °C
- Saturation concentration:40 g/m3 (20 °C) Air
- Hazard Class:3
- Hazard Class Description:Flammable liquid
:
:106.17 g/mol
:13,03801
:136.2 °C (1013 hPa)
:–95 °C
:0.867 g/cm³ (20 °C)
:15 °C
:Ambient
:202-849-4
:1175
:3,
:M-Clarity™ quality level = MQ100
:GHS02
:GHS08
:GHS07
:100-41-4
:C₈H₁₀
:MFCD00011647