Centrifuge tubes, conical bottom
Supplier: Simport Scientific
Certificates
About this item
Tubes made of either polystyrene, optically clear or polypropylene, translucent.
- Molded graduations from 2,5 to 50 ml
- Polypropylene tubes can be autoclaved
- Sterile or non sterile available
These centrifuge tubes are also useful for collecting and transporting biological specimens. Leakproof characteristics are ensured by a flat top plastic screw cap with an inner sealing lip.
Ordering information: Tubes are available in bags or in polypropylene racks for better protection during transport, storage and for convenient laboratory use. Racks can hold up to 25 tubes.
Delivery: Tubes are supplied sterile with green caps or non sterile with yellow caps.